Would you like to log in again?
Forceps, Iris Tissue, Straight, Serrated, Mortech
Supplier: Mortech
AB035
76548-692EA 23.34 USD InStock
76548-692
Forceps, Iris Tissue, Straight, Serrated, Mortech
Forceps
Serrated tip forceps suitable for delicate post-mortem dissection.
  • Standard grade stainless steel
  • Serrated pattern tips
  • Ribbed grip
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty