Would you like to log in again?
Forceps, Iris Tissue, Straight 1×2 Teeth, Mortech
Supplier: Mortech
AB037
76548-696EA 18.75 USD InStock
76548-696
Forceps, Iris Tissue, Straight 1×2 Teeth, Mortech
Forceps
Sharp 1×2 interlocking teeth for secure grip with minimal tissue compression.
  • Straight tips with 1×2 teeth
  • Standard grade stainless steel
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty