Would you like to log in again?
Forceps, Specimen Paddle, Mortech
Supplier: Mortech
AB043 AB064 AB042
76548-746EA 101.5 USD InStock
76548-746 76548-708 76548-706
Forceps, Specimen Paddle, Mortech
Forceps
Paddle forceps hold all types of tissue with the secure grip needed to accomplish thin sections for analysis. Thin sections are essential for proper infiltration of reagents prior to study. Great for training residents and technicians on thin section technique, cutting thin sections of fatty tissue like breast tissue and cutting even thickness sections of any type tissue.
  • Standard grade stainless steel
  • Ribbed grip
  • Paddle ends
Available in a variety of paddle types.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty