Would you like to log in again?
Forceps, Delicate Tissue, Curved Tip, Mortech
Supplier: Mortech
AB145 AB045
76548-712EA 47.85 USD InStock
76548-712 76548-848
Forceps, Delicate Tissue, Curved Tip, Mortech
Forceps
Delicate curved tips ideal for soft tissue specimens.
  • Stainless steel
  • Smooth grip
  • Curved tip
Available in standard or premium grade.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty