Would you like to log in again?
Forceps, Bayonet, Mortech
Supplier: Mortech
AB050
76548-722EA 109.14 USD InStock
76548-722
Forceps, Bayonet, Mortech
Forceps
Ergonomically designed for a more comfortable wrist position during post-mortem dissections. Offers speculum function for hollow cavity specimens such as uterus, bladder and colon.
  • Standard grade stainless steel
  • Off-set serrated tip
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty