Would you like to log in again?
Forceps, Ergonomic, Angled Tip, Mortech
Supplier: Mortech
AB055
76548-730EA 101.5 USD InStock
76548-730
Forceps, Ergonomic, Angled Tip, Mortech
Forceps
Unique perforated paddle handles for secure slip-free grip.
  • Stainless steel
  • Broad grip
  • Angled tip
Fingertip aligned perforations offer precise maneuvering. Extremely lightweight and versatile for a wide variety of tissue specimens. Can be cleaned with any laboratory cleaner or disinfectant.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty