Would you like to log in again?
Forceps, Rubber Tipped, Mortech
Supplier: Mortech
AB060 AB063 AB061 AB062
76548-744EA 35.61 USD InStock
76548-744 76548-740 76548-742 76548-738
Forceps, Rubber Tipped, Mortech
Forceps
Soft rubber tipped forceps allow for secure post-mortem dissection with minimal tissue damage or compression. An excellent option for techniques that require no metal exposure. Blue tips provide excellent contrast for specimen photography.
  • Standard grade stainless steel
  • Rubber rounded tips
Available in a variety of lengths.
NOTE: Do not autoclave. Rinse under water or wipe clean with any of our disinfecting cleaners.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty