Would you like to log in again?
Forceps, CutMATE Starter Kit, Mortech
Supplier: Mortech
AB070
76548-748EA 2235.55 USD InStock
76548-748
Forceps, CutMATE Starter Kit, Mortech
Forceps
CutMATE forceps were developed to overcome the variability of tissue specimen thickness. They will allow cutting consistent tissue specimen thickness of 2, 3, or 4 mm.
  • 304 stainless steel
The unique 'trident-like' design of the 35 mm long ergonomically angled prongs, ensures uniform tissue thickness cutting of tissue lengths that will fit standard cassettes. The inside surface of the prongs is specially textured to provide the extra grip needed for stabilizing and slicing difficult and hard to hold specimens.
Delivery information: Includes (4) forceps, decontamination rack, and calibration device.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty