Would you like to log in again?
Forceps, ProCUT Kit, Mortech
Supplier: Mortech
AB079
76548-766EA 1578.34 USD InStock
76548-766
Forceps, ProCUT Kit, Mortech
Forceps
ProCUT is an innovative tool that assists in cutting in parallel slices of uniform thickness.
  • Laser shaped AISI 304 stainless steel
  • 5 mm thickness slice
  • PTFE positioning base
ProCUT eliminates the guesswork in cutting a specific thickness of tissue mass. Suitable for use with both conventional and microwave tissue processors. Serial wet tissue slices are taken in a simple step with the unique design of ProCUT. Movable forceps prongs designed for ergonomic handling of varying sized tissues. Metal side plates secure sample in position. No more complaints about slicing being “too thick” or “not parallel”.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty