Would you like to log in again?
Forceps, Delicate, Extra Fine Sharp Tips, Mortech
Supplier: Mortech
AB132
76548-836EA 86.15 USD InStock
76548-836
Forceps, Delicate, Extra Fine Sharp Tips, Mortech
Forceps
Extra fine tipped forceps designed for the most delicate post-mortem dissections.
  • Premium grade stainless steel
  • Smooth grip
  • Extra fine sharp tips
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty