Would you like to log in again?
Forceps, Flat Angle Tip, Delicate, Mortech
Supplier: Mortech
AB133
76548-838EA 103.02 USD InStock
76548-838
Forceps, Flat Angle Tip, Delicate, Mortech
Forceps
Raised angled flat platform tip allows for grasping the finest tissue with a firm but controlled grip.
  • Premium grade stainless steel
  • Angle tips
  • Smooth grip
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty