Would you like to log in again?
Forceps, Adson Dressing, Tungsten Carbide, Mortech
Supplier: Mortech
AB402 AB403
76548-876EA 328.2 USD InStock
76548-876 76548-878
Forceps, Adson Dressing, Tungsten Carbide, Mortech
Forceps
Tungsten carbide tipped straight forceps with very fine, serrated tips for a wide variety of post-mortem dissection uses.
  • Premium grade tungsten carbide
  • Serrated, fine tips
  • Broad ribbed grip
Available with or without teeth.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty