Would you like to log in again?
Forceps, Precision Tipped, Insert, Disposable, Mortech
Supplier: Mortech
AB605
76548-892EA 55.52 USD InStock
76548-892
Forceps, Precision Tipped, Insert, Disposable, Mortech
Forceps
Disposable precision tipped fine toothed forceps offer a safe and exposure free alternative.
  • Stainless steel
  • Precision insert tip
  • Pack of 12
Tip inserts are designed for easy removal and disposal.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty