Would you like to log in again?
Aunex Scalpel Blade Remover, Mortech
Supplier: Mortech
ABR-100U
76548-896EA 43.28 USD InStock
76548-896
Aunex Scalpel Blade Remover, Mortech
Blade Removers and Dispensers
The Aunex ABR-100U scalpel blade remover provides a simply safer way of removing scalpel blades. Remove and dispose of used blades with the single-handed blade removal ABR-100U technique. No fingers, forceps, or re-sheathing. No Injuries.
  • Simply push and pull to remove scalpel blades without forceps or fingers
  • First blade remover to remove larger pathology and autopsy blades
  • Removes #40, #60, and #70 blades, in addition to standard blades size 6 to 26
  • Remove and dispose of up to 200 blades
The Aunex blade remover deploys the innovative patent pending Uniflex technology that simply lifts and traps the heel of a blade, then safely disengages and removes the blade. No moving parts. No jamming gears or mechanism.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty