Would you like to log in again?
Knife Sharpener, Variable Speed, Mortech
Supplier: Mortech
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AH032 AH033
76549-334EA 2826.9 USD InStock
76549-334 76549-332
Knife Sharpener, Variable Speed, Mortech
Knives
This variable speed sharpener creates a precise angle and a consistent, exact bevel by sharpening, thinning, and honing which results in a stronger cutting edge. The honing wheels are mounted on parallel shafts that rotate in opposite directions.
  • Stainless steel case
Available in US and International voltage.
Ordering information: Replacement honing wheels available (See model AH034).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty