Would you like to log in again?
Knife, German Dissecting, Mortech
Supplier: Mortech
AH101
76549-354EA 433.95 USD InStock
76549-354
Knife, German Dissecting, Mortech
Knives
Lightweight and perfectly balanced post-mortem dissection knife with a hollow ground cutting edge.
  • Premium grade stainless steel
Handle has a smooth top edge for comfort and blade is hardened to withstand continuous sharpening.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty