Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Scalpel Blades, Nonsterile, Mortech
Supplier: Mortech
AJ143 AJ144 AJ153 AJ145 AJ119 AJ155 AJ141 AJ146 AJ154
76549-456EA 101.5 USD InStock
76549-456 76549-454 76549-448 76549-416 76549-468 76549-458 76549-470 76549-452 76549-472
Scalpel Blades, Nonsterile, Mortech
Scalpels and Scalpel Assemblies Scalpel Blades
These nonsterile blades come individually wrapped and bulk boxed.
  • Nonsterile carbon steel
  • Pack of 100
  • Individually wrapped
Available in a variety of sizes and grades.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty