Would you like to log in again?
Qlicksmart® Blade Remover, BladeFLASK, Mortech
Supplier: Mortech
Qlicksmart®
AJ150
76549-464EA 55.52 USD InStock
76549-464
Qlicksmart® Blade Remover, BladeFLASK, Mortech
Blade Removers and Dispensers
The Qlicksmart® Blade Removal System is a medical safety device specially designed, engineered and manufactured to comply with recently revised OSHA Bloodborne Pathogens Standards. Qlicksmart® offers single-handed removal and immediate containment of contaminated scalpel blades which dramatically reduces the risk and cost of injury to the user.
  • Holds 100 blades
  • Fits blade nos. 10, 10a, 11, 12, 15, 20, 21, 22, 23, 24
  • Fits handle nos. 3 (AJ140), 4 (AJ101), 4L (AJ102), 5, 7, 9
  • Easy mounting with optional AJ151 mounting bracket
  • OSHA compliant
Features a built-in counter and automatic shut off when full.
Accessories information: The mounting bracket is necessary for the Qlicksmart® Blade Removal System to comply with recently revised OSHA Bloodborne Pathogens Standards. Qlicksmart® offers single-handed removal and immediate containment of contaminated scalpel blades which dramatically reduces the risk and cost of injury to the user when properly mounted.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty