Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Caliper, Analog, Mortech
Supplier: Mortech
BA040
76549-516EA 547.3 USD InStock
76549-516
Caliper, Analog, Mortech
Calipers
Excellent for measuring all tissue specimens with a sliding caliper and a finger tip nut to lock down ruler. Tool has graduated etched standard and metric markings.
  • Opens to 3" (76 mm)
  • Premium grade stainless steel
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty