Would you like to log in again?
Mopec Safe Seal System, Pre-Cut Bags Starter Pack, Mortech
Supplier: Mortech
BE160
76549-648EA 2736.51 USD InStock
76549-648
Mopec Safe Seal System, Pre-Cut Bags Starter Pack, Mortech
Bags Storage Bags
Mopec Safe Seal System is a military tested, heavy duty, hermetically sealable Bio-Safety Level 4 (BSL-4) Containment body bag system.
It is easy to use, as the body is placed in the flexible material, then heat-sealed with the Safe Seal Heat Sealer to provide complete and secure containment of infectious human or animal remains. The Safe Seal System was designed to help keep workers and survivors safe after a mass fatality event or any high-risk biohazard exposure.
Delivery information: The Starter Pack contains 25 pre-cut 48×96" bags (BE161), 1 heat sealer (BE153), and 2 alignment clips (BE900).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty