Would you like to log in again?
Body Bag, USA Response, Standard Duty, Envelope Zipper, Mortech
Supplier: Mortech
BE173
76549-664EA 61.64 USD InStock
76549-664
Body Bag, USA Response, Standard Duty, Envelope Zipper, Mortech
Bags Storage Bags
Mopec has partnered with multiple vinyl and polyethylene manufacturers to meet the demand for body bags.
  • 7-12 Mil PVC
  • Leak proof: seams are 100% RF heat welded/sealed
  • Envelope zipper
  • No Handles
  • Weight capacity 200 pounds
  • Color varies
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty