Would you like to log in again?
Body Bag, Poly, Standard Duty, Center Zipper, No Handles, Mortech
Supplier: Mortech
BE176
76549-668EA 23.34 USD InStock
76549-668
Body Bag, Poly, Standard Duty, Center Zipper, No Handles, Mortech
Bags Storage Bags
Made from enforced polyethylene, this standard duty body bag features a center zipper, no handles, and is suitable for cremation.
  • Adult size
  • Enforced coextruded polyethylene
  • Standard duty weight class
  • Straight center zipper
  • No handles
  • 8 Mil thick
  • Suitable for cremation
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty