Would you like to log in again?
VWR® Hotplates
Supplier: VWR
VWR®
H3880-H-VWR H3710-H-VWR
76549-922EA 716.49 USD InStock
76549-922 76549-914
VWR® Hotplates
Heaters Hot Plates
VWR® hotplates with temperature control up to 400 °C.
  • White ceramic surface for excellent chemical resistance
  • Digital speed control up to 400 °C
  • Available with external temperature probe (displays actual sample temperature)
  • 'HOT' LED safety indicator alerts user when plate is unsafe to touch
Corresponding magnetic hotplate stirrers (heating and stirring 76549-912, 76549-918, 76549-920 and 76549-926) or magnetic stirrers (stirring only 76549-916 and 76549-924) with ceramic plates are also available. Full specification details are given under separate entries, please view full details by searching for the relevant product code.
Certifications: CE marked.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty