Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Magnetic Hotplate Stirrers
Supplier: VWR
VWR®
H3710-HS-P1-VWR H3880-HS-VWR H3880-HS-P1-VWR H3710-HS-VWR
76549-920EA 939.32 USD InStock
76549-920 76549-926 76549-912 76549-918
VWR® Magnetic Hotplate Stirrers
Stirrers Magnetic Hotplate Stirrers
VWR® magnetic hotplate stirrers, with temperature control up to 400 °C and stirring speeds up to 1500 rpm.
  • White ceramic surface for excellent chemical resistance
  • Digital speed control up to 400 °C
  • Available with external temperature probe (displays actual sample temperature)
  • 'HOT' LED safety indicator alerts user when plate is unsafe to touch
Corresponding magnetic stirrers (stirring only 76549-916 and 76549-924) or hotplates (heating only 76549-914 and76549-922) with ceramic plates are also available. Full specification details are given under separate entries, please view full details by searching for the relevant product code.
Certifications: CE marked.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty