Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Vortex Mixer
Supplier: Avantor
VWR®
BV1000-GM-VWR
76549-928EA 284.52 USD InStock
76549-928
VWR® Vortex Mixer
Shakers and Mixers Vortex Mixers
VWR® variable speed vortex mixer with standard cup head.
  • High powered motor drive for instant vortexing
  • Adjustable speed control
  • Optional tube holders for hands-free mixing
VWR® vortex mixer provides variable speed control up to 3200 rpm with touch activation.
Certifications: CE marked
Delivery information: Supplied with standard cup head, a range of optional accessories are also available to purchase separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty