Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Mini Vortex Mixer

Supplier: Avantor
VWR®
BV205-VWR
76549-932EA 198.2 USD InStock
76549-932
VWR® Mini Vortex Mixer
Shakers and Mixers Vortex Mixers
VWR® mini vortex mixer with standard cup head.

  • Extremely compact design
  • Powerful mixing for tubes up to 50 ml
  • One handed, pressure activated operation

VWR® mini vortex mixer provides high speed vortexing (2750 rpm) with touch activation.

Certifications: CE marked

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty