Would you like to log in again?
High Performance Muffle Furnace, FP Series, Yamato
Supplier: Yamato
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FP-313 FP-103 FP-413 FP-303
76550-238EA 9240.95 USD InStock
76550-238 76550-240 76550-242 76550-244
High Performance Muffle Furnace, FP Series, Yamato
Ovens Furnaces (Ashing and Muffle Ovens)
Muffle furnace is designed to maintain uniform high heat temperatures and make the unit last for years. The units in this line have firmly sealed chamber doors to provide excellent heat tightness. They also come with a top-quality alumina porcelain hot plate where the heater is not exposed to the inner chamber. This ensures that no sample contamination will take place.
  • RCF-free chamber
  • High-temperature accuracy at ±1.0 °C
  • Upgraded with long-life R-thermocouple sensors
  • High accuracy controller for better operability and visibility
  • Excellent heat tightness with a firmly sealed chamber door
  • Program operation of maximum 99 steps, 99 patterns, with repeat operation function
High-quality alumina porcelain hot plate where the heater is not exposed to the inner chamber preventing contamination of samples.
Safety features include self-diagnostic functions, calibration off-set, key lock function, auto-recovery after a power failure, earth leakage breaker, automatic overheat prevention device, and independent overheat prevention device.
Optional items include exhaust system, N₂ gas loading device (with flow meter), temperature output terminal, time-up output terminal, sample tray, event output terminal, operation signal output terminal and furnace floor plate.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty