Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnetic Stirrers with Hotplate, 220 V

Supplier: Yamato
MH-520-220V MH-301-220V MH-800-220V MG-600H-220V
76550-260EA 1949.41 USD InStock
76550-260 76550-252 76550-264 76550-256
Magnetic Stirrers with Hotplate, 220 V
Stirrers Magnetic Hotplate Stirrers
These magnetic stirrers include ceramic-coated aluminum plates.

  • 220 V model
  • Stirring rate up to 1500 rpm
  • Stirring capacity up to 10000 ml
  • Chemical-Resistant plate
  • Triac input temperature control

The plates are chemical resistant and heat conductive.

Each magnetic stirrer includes a stirrer bar.

Caution: 220 V models. Corresponding model with voltage suitable for use in US (76550-250, 76550-254, 76550-258, 76550-262) are also available, please see separate product page.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty