Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® C-Leg Lab Benches, Frame + Top + Uprights + Shelves, Hand Crank, Snow White

Supplier: Avantor

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
14-C101SW429 14-C101SW420 14-C101SW433 14-C101SW468 14-C101SW428 14-C101SW437 14-C101SW464 14-C101SW469 14-C101SW425 14-C101SW438 14-C101SW435 14-C101SW467 14-C101SW434 14-C101SW465 14-C101SW430 14-C101SW466 14-C101SW474 14-C101SW471 14-C101SW423 14-C101SW457 14-C101SW426 14-C101SW463 14-C101SW421 14-C101SW439 14-C101SW473 14-C101SW470 14-C101SW436 14-C101SW431 14-C101SW458 14-C101SW427 14-C101SW424 14-C101SW432 14-C101SW462 14-C101SW459 14-C101SW422 14-C101SW472 14-C101SW460 14-C101SW456 14-C101SW461 14-C101SW475
76550-292EA 3939.21 USD InStock
76550-292 76550-826 76551-018 76550-906 76550-334 76550-412 76550-478 76550-578 76550-952 76550-634 76550-436 76550-656 76550-876 76550-712 76550-976 76550-614 76550-658 76550-814 76551-192 76550-350 76550-374 76550-770 76550-672 76550-310 76550-992 76550-498 76551-000 76551-040 76550-786 76550-842 76550-668 76550-624 76550-528 76551-080 76550-660 76550-782 76550-486 76550-882 76550-762 76550-322
VWR® C-Leg Lab Benches, Frame + Top + Uprights + Shelves, Hand Crank, Snow White
Furniture Tables and Workbenches
VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel. There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality.

  • Snow white frame with hand crank style
  • Ergonomic and highly adaptable to the needs of the user
  • Flexible, modular design
  • Each shelf measures 11.81" in depth
  • Offering a wide range of accessories to meet any need
  • Various concept workbench sizes and combinations
  • Impressive stability with its steel frame and maximum load capacity of 1100 lbs.
  • Cleanroom suitability ISO class 7

The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. VWR® lab benches are height adjustable in manual, motorized and hand crank versions. All bench frames can be equipped with casters to create a mobile workstation solution.

Height adjustment available either electrically (electric motor height adjustment) or with a hand crank or an allen key. A wide range of accessories and add-ons, making concept workbenches easy to customize.

Accessories information: Upright accessories can be added in any position along the upright in 1" increments. Some accessories are bench style or size dependent.

Ordering information: Manual, hand crank, and motorized frame styles are available, as well as several configurations, including frame only, frame + top, frame + top + uprights, frame + top + uprights + shelves, and frames with notches for uprights (all sold separately). Contact your Avantor representative for more information. Notched work surfaces add 2.625" to depth of table frame. Work surfaces for Single bay uprights include 2 notches (left and right); work surfaces for double bay uprights include 3 notches (left, center, right). Tables ship LTL, flat-packed. Installation not included.

Packaging: Benches are shipped knocked down, on non-stackable pallets. Assembly instructions and hardware included.

Order Now
 

ORDER

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty