Would you like to log in again?
VWR® Reagent Rack Island Shelving System
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
14-9566154 14-9566146 860095-VWR 14-9566155 14-9566156 14-956SW54 860099-SW 14-9566149 14-9566141 14-9566111 14-9566148 14-9566157 14-956SW59 14-9566110 14-956SW56 14-9566112 14-9566143 14-956SW58 14-9566158 860098-VWR 14-9566152 14-956SW53 14-956SW55 14-9566153 14-9566150 14-9566147 14-9566144 860096-SW 14-9566161 14-9566160 14-956SW60 860097-VWR 14-956SW52 14-956SW61 14-9566108 14-9566142 14-9566151 14-9566140 860096-VWR 14-956SW57 14-9566145 860099-VWR 14-9566159 14-9566109 14-9566139 14-9566138 860094-VWR
10808-890EA 856.62 USD InStock
10808-890 10808-870 10808-892 10808-894 10808-850 10808-872 10808-852 10808-830 10808-896 10808-874 10808-832 10808-854 10808-876 10808-856 10808-878 10808-834 10808-858 10808-836 76550-434 76550-516 76550-650 76550-410 76551-064 10808-880 10808-882 76551-084 10808-860 10808-840 76550-618 10808-884 10808-862 76551-106 10808-864 10808-842 10808-886 10808-888 10808-844 10808-866 76551-028 10808-868 10808-846 10808-848 10808-826 10808-828 76550-504 76550-460 76550-540
VWR® Reagent Rack Island Shelving System
Racks Tube Racks
VWR® Reagent Rack systems provide excellent storage of laboratory equipment above the lab station.
  • Sturdy shelving for solid storage
  • Variety of heights available
These systems are an ideal addition to new or existing laboratory workspaces and casework, where additional shelving is needed
Uprights include attached brackets. Single shelf bays include two uprights with left/right facing brackets. Extensions include a single upright with a center oriented bracket. When an extension is added to a single shelf bay, the extension upright becomes the middle upright in the system.
The VWR® Reagent Rack uprights are available in three heights and can support a range of shelf and power options. Standard VWR® slotted upright shelves and power rails can be mounted to this island shelving system.
Ordering information: Single Shelf Bays include two uprights with attached brackets, one top cross rail, and two corner brackets. Extensions include one upright with attached bracket, one top cross rail, and one "T" bracket. Shelves must be ordered separately.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty