Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® C-Leg Lab Benches, Frame + Top + Uprights, Snow White

Supplier: Avantor

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
14-C100SW449 14-C101SW414 14-C100SW413 14-C100SW397 14-C103SW413 14-C100SW398 14-C101SW455 14-C103SW396 14-C103SW446 14-C103SW447 14-C103SW415 14-C100SW393 14-C101SW449 14-C100SW447 14-C101SW393 14-C103SW395 14-C101SW413 14-C100SW446 14-C103SW453 14-C103SW449 14-C101SW452 14-C100SW416 14-C101SW454 14-C103SW448 14-C101SW448 14-C100SW455 14-C100SW453 14-C100SW396 14-C100SW448 14-C101SW415 14-C103SW398 14-C101SW398 14-C103SW451 14-C103SW394 14-C101SW447 14-C101SW450 14-C100SW450 14-C103SW397 14-C100SW395 14-C100SW451 14-C103SW393 14-C101SW395 14-C103SW454 14-C100SW454 14-C101SW446 14-C100SW414 14-C103SW416 14-C103SW450 14-C101SW451 14-C100SW415 14-C100SW394 14-C101SW396 14-C101SW394 14-C103SW414 14-C103SW452 14-C101SW416 14-C101SW397 14-C100SW452 14-C101SW453 14-C103SW455
76551-034EA 2897.21 USD InStock
76551-034 76551-076 76551-198 76550-390 76550-948 76550-828 76550-796 76550-312 76550-676 76550-830 76550-358 76550-556 76550-678 76550-956 76550-670 76550-990 76550-354 76550-476 76550-630 76551-046 76551-164 76551-086 76551-160 76550-840 76550-720 76550-368 76550-524 76550-766 76550-724 76550-802 76550-482 76550-362 76550-484 76550-320 76550-640 76551-094 76550-728 76551-016 76551-136 76550-774 76550-336 76550-856 76550-418 76550-690 76550-332 76550-970 76550-574 76550-894 76551-066 76550-666 76550-468 76550-942 76550-426 76550-988 76550-944 76550-506 76550-384 76550-462 76550-860 76550-586
VWR® C-Leg Lab Benches, Frame + Top + Uprights, Snow White
Furniture Tables and Workbenches
VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel. There are many possible adjustments, all components fit together seamlessly and each detail is designed for real world functionality.

  • Snow white frame
  • Ergonomic and highly adaptable to the needs of the user
  • Flexible, modular design
  • Offering a wide range of accessories to meet any need
  • Various concept workbench sizes and combinations
  • Height adjustment available either electrically (electric motor height adjustment) or with a hand crank or an Allen key
  • A wide range of accessories and add-ons, making concept workbenches easy to customize
  • Impressive stability with its steel frame and maximum load capacity of 1100 lbs.
  • Cleanroom suitability ISO class 7

The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. VWR® lab benches are height adjustable in manual, motorized and hand crank versions. All bench frames can be equipped with casters to create a mobile workstation solution.

Accessories information: Upright accessories can be added in any position along the upright in 1" increments. Some accessories are bench style or size dependent.

Ordering information: Shelves sold separately. Manual, hand crank, and motorized frame styles are available, as well as several configurations, including frame only, frame + top, frame + top + uprights, frame + top + uprights + shelves, and frames with notches for uprights (all sold separately). Contact your Avantor representative for more information.
Notched work surfaces add 2.625" to depth of table frame. Work surfaces for single bay uprights include two notches (left and right); work surfaces for double bay uprights include three notches (left, center, right).
Tables ship LTL, flat-packed. Installation not included.

Packaging: Benches are shipped knocked down, on non-stackable pallets. Assembly instructions and hardware included.

Order Now
 

ORDER

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty