Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® C-Leg Lab Benches, Frames with Notches for Uprights, Snow White

Supplier: Avantor

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
14-C100SW490 14-C100SW492 14-C103SW487 14-C100SW488 14-C100SW486 14-C101SW496 14-C101SW482 14-C101SW495 14-C103SW494 14-C103SW489 14-C101SW492 14-C100SW499 14-C103SW488 14-C100SW482 14-C103SW482 14-C101SW488 14-C101SW489 14-C100SW498 14-C101SW498 14-C103SW486 14-C100SW489 14-C101SW494 14-C103SW484 14-C100SW485 14-C100SW483 14-C100SW484 14-C103SW493 14-C101SW481 14-C100SW495 14-C100SW491 14-C101SW483 14-C103SW480 14-C101SW493 14-C100SW494 14-C103SW497 14-C103SW492 14-C100SW496 14-C100SW497 14-C101SW490 14-C101SW491 14-C100SW481 14-C100SW493 14-C100SW487 14-C101SW485 14-C103SW495 14-C101SW497 14-C103SW490 14-C103SW481 14-C101SW480 14-C103SW499 14-C103SW483 14-C101SW484 14-C101SW486 14-C103SW485 14-C103SW496 14-C103SW498 14-C101SW487 14-C100SW480 14-C101SW499 14-C103SW491
76551-032EA 4634.35 USD InStock
76551-032 76551-110 76551-030 76550-706 76551-116 76551-114 76550-554 76550-512 76550-756 76550-638 76550-394 76550-550 76550-430 76550-750 76551-088 76550-716 76551-048 76550-444 76550-644 76550-568 76550-888 76550-646 76550-844 76550-768 76550-648 76550-680 76550-560 76550-682 76550-562 76551-010 76551-132 76551-174 76550-408 76550-928 76550-610 76550-974 76550-338 76550-538 76550-858 76550-496 76550-850 76551-068 76551-022 76551-104 76551-026 76550-422 76550-940 76550-984 76550-302 76550-424 76550-702 76550-746 76550-548 76550-824 76550-704 76550-580 76550-386 76550-982 76550-300 76550-388
VWR® C-Leg Lab Benches, Frames with Notches for Uprights, Snow White
Furniture Tables and Workbenches
VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel. There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality.

  • Snow white frame
  • Ergonomic and highly adaptable to the needs of the user
  • Flexible, modular design
  • Offering a wide range of accessories to meet any need
  • Various concept workbench sizes and combinations
  • Height adjustment available either electrically (electric motor height adjustment) or with a hand crank or an Allen key
  • A wide range of accessories and add-ons, making concept workbenches easy to customize
  • Impressive stability with its steel frame and maximum load capacity of 1100 lbs.
  • Cleanroom suitability ISO class 7

The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. VWR® lab benches are height adjustable in manual, motorized and hand crank versions. All bench frames can be equipped with casters to create a mobile workstation solution.

Accessories information: Upright accessories can be added in any position along the upright in 1" increments. Some accessories are bench style or size dependent.

Ordering information: Uprights and shelves are sold separately. Manual, hand crank and motorized frame styles are available, as well as several configurations, including frame only, frame + top, frame + top + uprights, frame + top + uprights + shelves and frames with notches for uprights (all sold separately). Contact your Avantor representative for more information.
Notched work surfaces add 2.625" to depth of table frame. Work surfaces for single bay uprights include two notches (left and right); work surfaces for double bay uprights include three notches (left, center, right).
Tables ship LTL, flat-packed. Installation not included.

Packaging: Benches are shipped knocked down, on non-stackable pallets. Assembly instructions and hardware included.

Order Now
 

ORDER

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty