Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® C-Leg Lab Benches, Frame + Top, Snow White

Supplier: Avantor

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
14-C100SW403 14-C103SW444 14-C101SW124 14-C103SW440 14-C103SW442 14-C103SW403 14-C103SW441 14-C103SW445 14-C101SW440 14-C100SW442 14-C101SW123 14-C101SW443 14-C100SW444 14-C101SW121 14-C101SW403 14-C101SW125 14-C100SW441 14-C103SW125 14-C103SW443 14-C100SW122 14-C103SW123 14-C100SW445 14-C100SW440 14-C101SW442 14-C101SW445 14-C101SW441 14-C100SW121 14-C103SW121 14-C100SW123 14-C103SW122 14-C101SW444 14-C100SW443 14-C100SW124 14-C101SW122 14-C103SW124 14-C100SW125
76551-056EA 3876.59 USD InStock
76551-056 76551-156 76551-098 76550-628 76550-730 76550-378 76550-896 76550-314 76550-612 76550-810 76550-438 76550-912 76550-392 76551-092 76550-372 76551-090 76550-790 76550-296 76550-450 76551-190 76551-100 76550-958 76550-838 76550-938 76551-204 76550-742 76550-446 76550-402 76550-986 76550-964 76550-920 76550-404 76550-822 76551-180 76550-440 76550-542
VWR® C-Leg Lab Benches, Frame + Top, Snow White
Furniture Tables and Workbenches
VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel. There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality.

  • Snow white frame
  • Ergonomic and highly adaptable to the needs of the user
  • Flexible, modular design
  • Offering a wide range of accessories to meet any need
  • Various concept workbench sizes and combinations
  • Height adjustment available either electrically (electric motor height adjustment) or with a hand crank or an allen key
  • A wide range of accessories and add-ons, making concept workbenches easy to customize
  • Impressive stability with its steel frame and maximum load capacity of 1100 lbs.
  • Cleanroom suitability ISO class 7

The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. VWR® lab benches are height adjustable in manual, motorized and hand crank versions. All bench frames can be equipped with casters to create a mobile workstation solution.

Accessories information: Upright accessories can be added in any position along the upright in 1" increments. Some accessories are bench style or size dependent.

Ordering information: Uprights and shelves are sold separately. Manual, hand crank, and motorized frame styles are available, as well as several configurations, including frame only, frame + top, frame + top + uprights, frame + top + uprights + shelves, and frames with notches for uprights (all sold separately). Contact your Avantor representative for more information. Frames with notches for uprights (sold separately) are required for upright and shelf accessories. Notched work surfaces add 2.625" to depth of table frame. Work surfaces for single bay uprights include 2 notches (left and right); work surfaces for double bay uprights include 3 notches (left, center, right). Tables ship LTL, flat-packed. Installation not included.

Packaging: Benches are shipped knocked down, on non-stackable pallets. Assembly instructions and hardware included.

Order Now
 

ORDER

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty