Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® 4-Leg Lab Benches, Snow White

Supplier: Avantor

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
14-CS917-SW 14-CS916-SW 14-CS909-SW 14-CS932-SW 14-CS905-SW 14-CS918-SW 14-CS943-SW 14-CS928-SW 14-CS901-SW 14-CS911-SW 14-CS926-SW CS712-SW 14-CS935-SW 14-CS931-SW 14-CS907-SW 14-CS920-SW 14-CS910-SW 14-CS946-SW 14-CS921-SW 14-CS912-SW 14-CS904-SW 14-CS944-SW 14-CS936-SW 14-CS914-SW 14-CS947-SW 14-CS940-SW 14-CS925-SW 14-CS930-SW 14-CS906-SW 14-CS903-SW 14-CS922-SW 14-CS908-SW 14-CS933-SW 14-CS949-SW 14-CS915-SW 14-CS924-SW 14-CS941-SW 14-CS919-SW 14-CS945-SW 14-CS942-SW 14-CS902-SW 14-CS927-SW 14-CS948-SW 14-CS929-SW 14-CS913-SW 14-CS934-SW CS718-SW CS715-SW 14-CS923-SW
76551-112EA 2597.99 USD InStock
76551-112 76550-508 76550-908 76550-798 76550-910 76550-878 76550-836 76550-590 76550-592 76550-352 76550-552 76550-794 76550-398 76550-872 76551-200 76550-916 76551-126 76550-686 76550-324 76550-522 76550-886 76550-846 76550-726 76551-082 76550-442 76550-520 76551-134 76551-052 76551-050 76550-490 76550-926 76551-058 76550-534 76550-930 76550-778 76550-536 76550-812 76550-978 76550-492 76550-330 76550-298 76550-530 76551-188 76551-020 76550-500 76550-588 76550-744 76550-626 76550-308
VWR® 4-Leg Lab Benches, Snow White
Furniture Tables and Workbenches
VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel.

  • Snow white frame
  • Legged steel structure-4
  • Load capacity 2000 lbs. for even the toughest environments
  • Can be easily equipped with accessories
  • ISO class 7 cleanroom compatible

There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality. The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. All bench frames can be equipped with casters to create a mobile workstation solution.

Our 4-leg frame height range can be adjusted from 27.56 - 40.55", in 1" increments, making these tables suitable for work while sitting or standing. Shelves depth 11.81".

Ordering information: Notched work surfaces add 2.625" to depth of table frame. Work surfaces for single bay uprights include two notches (left and right); work surfaces for double bay uprights include three notches (left, center, right).
Tables ship LTL, flat-packed. Installation not included.

Packaging: Benches are shipped knocked down, on non-stackable pallets. Assembly instructions and hardware included.

Order Now
 

ORDER

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty