Would you like to log in again?
PROPOR HC MURUS Capsules, Parker Bioscience
Supplier: Parker
ZLHC2-620BB-PSL ZLHC3-620BB-PNL ZLHC2-620BB-PNL ZLHC1-620BB-PSL ZLHCK-620BB-PNL ZLHCK-620BB-PSL ZLHC1-620BB-PNL ZLHC3-620BB-PSL
76552-174EA 1082.37 USD InStock
76552-174 76541-094 76541-096 76552-166 76552-170 76552-172 76541-090 76552-168
PROPOR HC MURUS Capsules, Parker Bioscience
Filters Capsule Filters
PROPOR HC sterilizing grade filters have been specifically designed for the effective and economical processing of difficult to filter solutions.
  • Optimized membrane configuration allows up to 10X the throughput compared to single layer membrane products
  • Integral prefilter layer can condense filter trains for greater processing economy
  • Incorporates a fully validated and integrity testable 0.2 micron membrane for assurance of sterility
  • Low binding for minimal product loss
  • Single-use Capsule format available with Pre-sterile version by Gamma-irradiation
The optimized PROPOR HC PES membrane configuration features a highly asymmetric membrane prefilter layer, which significantly extends throughput and prevents the problems associated with premature filter blockage with complex solutions. PROPOR HC filters are high capacity and fast flowing.
The PES membrane is inherently low binding, which minimizes product loss due to protein or preservative adsorption. The filters have low extractable levels and broad chemical compatibility.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty