Would you like to log in again?
ChromCap™ 100 HPLC Cap Assembly, Foxx Life Sciences
Supplier: Foxx Life Sciences
450-0322-FLS 450-0621-FLS 450-0321-FLS 450-0320-FLS 450-0623-FLS 450-0624-FLS 450-0622-FLS 450-0620-FLS
76552-482EA 149.06 USD InStock
76552-482 76552-494 76552-484 76552-496 76552-490 76552-492 76552-486 76552-488
ChromCap™ 100 HPLC Cap Assembly, Foxx Life Sciences
Closures Bottle Closures
Foxx Life Sciences ChromCap™ 100 used in upstream caps and bottle system, offer the safest and most ergonomic lab safety solution in the industry.
  • Patented VersaCap® technologyprovides the flexibility of interchangeable connectors
  • To prevent spills and vapors fromleaking, tube fittings secure tubes tocontainer to create a close-loopsystem
  • Plugs included for all unused ports
Foxx Life Sciences ChromCap™ 100 used in upstream caps and bottle system, offer the safest and most ergonomic lab safety solution in the industry. This system comes with patented VersaCap®, a Class VI PTFE manifold, chemically resistant viton o-ring. The sealed VersaCap® design that Foxx offers eliminates tubes twisting as you thread the cap and eliminates the tubes from sliding in and out of the container.
Each assembly comes with either a 38-430 mm cap or a patented GL 45 cap and manifold assembly, high quality fittings for 1×3.2 mm (¹/₈") Ext.Ø tubing, 1×1.6 mm (¹/₁₆") Ext.Ø tubing and 1×plugs of 6.4 mm (¹/₄") for blocking of unused ports.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty