Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Abbott™ BinaxNOW® Streptococcus Pneumoniae Antigen Card Test Kit and Control, Healthcare Products
Supplier: Healthcare Products
BinaxNOW®
MPC710-010 MPC710-000
76553-958EA 82.35 USD InStock
76553-958 76553-892
Abbott™ BinaxNOW® Streptococcus Pneumoniae Antigen Card Test Kit and Control, Healthcare Products
Assays Immunological Assays
BinaxNOW® S. pneumoniae antigen card is a rapid assay for the qualitative detection of S. pneumoniae antigen in the urine of patients with pneumonia and in the cerebral spinal fluid (CSF) of patients with meningitis.
It is intended to aid, in conjunction with culture and other methods, in the diagnosis of both pneumococcal pneumonia and pneumococcal meningitis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty