Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Abbott™ DIGIVAL™ BinaxNOW® Interface, Kits and Accessories for Influenza A&B, Healthcare Products
Supplier: Healthcare Products
BinaxNOW®
MPC575-000 MPCARFLUSTART MPCARFLU8PK MPCARFLURR MPCARFLUPRT MPCLFR-003 MPCLFR-124 MPCARFLUBD MPC575-080 MPCMCP1810395 MPCARFLUBDPRT MPCGP15
76553-902EA 3444.31 USD InStock
76553-902 76553-946 76553-914 76553-922 76553-966 76553-938 76553-894 76553-908 76553-888 76553-932 76553-886 76553-974
Abbott™ DIGIVAL™ BinaxNOW® Interface, Kits and Accessories for Influenza A&B, Healthcare Products
Assays Immunological Assays
DIGIVAL™ Interface reads and interprets BinaxNOW® lateral flow tests.
  • OBJECTIVITY - Eliminate operator subjectivity
  • CONNECTIVITY - Combine the control of a connected instrument with the speed of a rapid assay
  • ACCURACY - Reduced potential for error- Data input, Test interpretation and Result reporting
  • QUALITY - Ensure quality controls are tested with our QC lockout feature
  • SPEED - Read and interpret approved rapid assays in seconds
DIGIVAL™ accurately reads and interprets lateral flow tests in just seconds, delivering automated, objective results in any healthcare setting. It is a camera based instrument that will detect the presence of and identify a completed lateral flow assay, analyze the intensity of the test and control line and display the results (positive, negative or invalid) on the color touch screen.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty