Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Abbott™ BinaxNOW™ Legionella Urinary Antigen Card Test Kit and Control, Healthcare Products
Supplier: Healthcare Products
BinaxNOW®
MPC852-010 MPC852-000
76553-928EA 67.04 USD InStock
76553-928 76553-964
Abbott™ BinaxNOW™ Legionella Urinary Antigen Card Test Kit and Control, Healthcare Products
Assays Immunological Assays
BinaxNOW™ Legionella urinary antigen card is a rapid assay for the qualitative detection of Legionella pneumophila serogroup 1 antigen in urine samples from patients with symptoms of pneumonia.
It is intended to aid in the presumptive diagnosis of Legionnaires’ disease caused by L. pneumophila serogroup 1 in conjunction with culture and other methods.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty