Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Abbott™ BinaxNOW™ Malaria Test Kits (US and Global) and Control, Healthcare Products
Supplier: Healthcare Products
BinaxNOW®
MPC665-025 MPC665-000 MPC665-010
76553-918EA 555.93 USD InStock
76553-918 76553-954 76553-896
Abbott™ BinaxNOW™ Malaria Test Kits (US and Global) and Control, Healthcare Products
Assays Immunological Assays
The BinaxNOW™ malaria test is an invitro immunochromatographic assay for the qualitative detection of plasmodium antigens circulating in human venous and capillary EDTA whole blood of individuals with signs and symptoms of malarial infection.
  • Results seen in 15 minutes
  • Enables immediate treatment
  • Simple test using only 1 reagent
  • Requires a small blood sample easily collected by fingerstick
  • No need for skilled microscopist
  • Differentiated results allow diagnosis of potentially lethal P. falciparum infection
  • Easy to read, qualitative results
The test targets the histidine-rich protein II (HRPII) antigen specific to Plasmodium falciparum (P.f.) and a pan-malarial antigen common to all four malaria species capable of infecting humans - P. falciparum, P. vivax (P.v.), P. ovale (P.o.), and P. malariae (P.m.).
Intended to aid in the rapid diagnosis of human malaria infections and in the differential diagnosis of Plasmodium falciparum (P.f.) infections from other less virulent malarial infections. Negative results must be confirmed by thin / thick smear microscopy.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty