Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Abbott™ BinaxNOW™ COVID-19 Ag Cards, Control For Use With NAVICA™ Mobile App, Healthcare Products
Supplier: Healthcare Products
BinaxNOW®
MPC195-000 MPC190-010 MPC195-080
76553-956EA 447.32 USD InStock
76553-956 76553-916 76553-960
Abbott™ BinaxNOW™ COVID-19 Ag Cards, Control For Use With NAVICA™ Mobile App, Healthcare Products
Assays Immunological Assays
Professional use.
  • Cost-effective, high performing test designed for decentralized testing
  • Visually read results in 15 minutes - no instrument required
  • Simple test procedure using a direct nasal swab
  • Onboard extraction allows the swab to be directly inserted into the test card
  • Room temperature storage
The 15-minute BinaxNOW™ COVID-19 Ag card, a rapid antigen test, was designed alongside the revolutionary NAVICA™ digital system for testing in CLIA-waived settings like universities, offices, pharmacies, long term care facilities.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty