Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Abbott™ BinaxNOW™ Strep A Test Kit and Control, Healthcare Products
Supplier: Healthcare Products
BinaxNOW®
MPC730-025 MPC730-010
76553-906EA 88.68 USD InStock
76553-906 76553-972
Abbott™ BinaxNOW™ Strep A Test Kit and Control, Healthcare Products
Assays Immunological Assays
BinaxNOW™ Strep A Test utilizes a simple to use on board extraction system.
  • Rapid response - Yields results in 6 minutes.
  • Accurate - Treat and test with confidence.
  • Flexible - Results while patient waits.
  • Simplicity - Onboard extraction.
  • Effective - Enables prompt and appropriate antibiotic therapy, thus reducing the course of the illness, lessening the severity of symptoms, and reducing the chances of further spread or complications.
The BinaxNOW™ Strep A test is a rapid immunochromatographic assay for the qualitative detection of Streptococcus pyogenes Group A antigen from throat swab specimens to aid in the diagnosis of Group A pharyngitis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty