Would you like to log in again?
COSA™ CIP 90 Alkaline Cleaner
Supplier: Ecolab
COSA™
6301053 6301054 6301053
76554-194EA 597.63 USD InStock
76554-194 76554-196 CA76554-194
COSA™ CIP 90 Alkaline Cleaner
Cleaning Agents Detergents and Surface Cleansers
Cosa™ CIP 90 is a mildly alkaline cleaner.
  • Mild alkaline formula allows for dual use in CIP or manual applications
  • Powerful blend of surfactants and sequestrants for broad cleaning ability
  • Supported by a complete validation package including techniques for determining acceptable residual levels and analytical methods for residue quantitation
For use in CIP pharmaceutical and cosmetic processing equipment, COSA™ CIP 90 is a mildly alkaline, surfactant containing cleaner. The product has broad cleaning ability and is ideal for the removal of organic soils such as fat, oil, and oil-in-water emulsions in piping, mixers, fluid beds, tanks, and storage vessels. The near-neutral formula also allows for manual cleaning.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty