Would you like to log in again?
Klercide™ 70/30 IPA
Supplier: Ecolab
Klercide™
6600136 6600135 6600097 6600100 6600139 6600101
76547-876CS 277.66 USD InStock
76547-876 CA76717-440 76554-222 CA77322-218 76554-224 CA76717-436
Klercide™ 70/30 IPA
Cleaning Agents Disinfectants
Klercide™ 70/30 IPA is sterile 70% v/v Isopropyl alcohol blended with Water for Injection for daily disinfection of critical areas. Klercide™ 70/30 IPA is a one-step disinfectant IPA spray designed for disinfection of cleanroom environments, with trigger-spray format featuring the patented “bag-in-a-bottle” SteriShield™ Delivery System.
  • Efficacy - 0.2 micron-filtered 70% isopropyl alcohol solution blended with WFI (endotoxin level guaranteed to be <0.25 E.U./mL) to deliver Bactericidal, Fungicidal and Virucidal efficacy.
  • Daily Disinfection - 70/30 IPA in a user-friendly trigger-spray format, designed to streamline daily disinfection in critical processing applications including aseptic processing, RABs, product contact surfaces and other sterile areas.
  • Ideal for Sanitizing - For sanitizing of hard, non-porous, food contact and non-food contact surfaces.
  • Sterile Delivery System - Trigger-spray format features patented SteriShield™ Delivery System (SDS), providing sterility assurance of contents in bottle by forming a complete seal that prevents any air from (and microbial or chemical contaminants) being drawn back into the bottle.
  • Sterile Production - Ultrasonically sealed and gamma irradiated in a Grade C/ISO Class 5 cleanroom; supported with certificates of analysis and Certificates of Sterility with every batch.
  • Convenient Packaging - Available as either double or triple-bagged to streamline movement into high-grade areas, with packaging designed for ease-of-use with gloved hands.
For use in pharmaceutical cleanrooms and associated areas, Klercide™ 70/30 IPA can be used for disinfection purposes on cleanroom surfaces and equipment. Available in a ready-to-use format, Klercide 70/30 IPA contains 70% v/v Isopropyl Alcohol blended with Water for Injection.
The product is 0.2 micron filtered and double bagged in an ISO 5 cleanroom before being terminally sterilized by gamma irradiation. Endotoxin level is guaranteed to be less than 0.25EU/ml.
Certifications: EPA Registered Disinfectant, Sanitizer, Fungicide and Virucide.
EPA Reg. # 1677-249
DIN Registered, DIN: 02447789
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty