Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Klerwipe™ 70/30 IPA 100% polyester pouch wipes

Supplier: Ecolab

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Klerwipe™
6600577
76554-264CS 341.04 USD InStock
76554-264
Klerwipe™ 70/30 IPA 100% polyester pouch wipes
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Klerwipe™ 70/30 IPA are sterile, knitted 100% polyester wipes impregnated with 0.2 micronfiltered IPA blended with deionized water.

  • Consistent Cleaning: Pre-saturated with 0.2 micron-filtered 70% isopropyl alcohol solution blended with WFI (endotoxin level <1 EU/wipe)
  • Ready-to-Use Wipes: Pre-saturated wipes are ready to use and enable one-step daily cleaning, ideal for use in material transfer and residue removal protocols
  • 100% Polyester Substrate: Premium-grade, 100% knitted polyester wipes are low-linting and pre-laundered in an ISO Class 4 cleanroom to significantly reduce particles
  • Sterile Production: Ultrasonically sealed and gamma irradiated in a Grade C/ISO Class 5 cleanroom; supported with certificates of analysis and Certificates of Sterility with every batch
  • Convenient Packaging: Available in large pack size ideal for high sessional use
  • Pouch wipes are double bagged to streamline movement into high-grade areas, with Kleartear for easy opening with gloved hands

Streamline daily cleaning, including material transfers in lower grade areas and residue removal protocols, with the one-step simplicity of ready-to-use sterile wipes pre-saturated with 70/30 IPA blended with DI (Deionized Water).

The Klerwipe™ 70/30 IPA 100% polyester wipes blended with DI are an ideal solution for those looking for the quality of polyester in a larger pack size than other Klerwipe pouch wipes.

Basis weight: 120-140 g/m²

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty