Would you like to log in again?
InVivoPure™ pH 8.0 Dilution Buffer, BioXCell
Supplier: BioXCell
IP0080-50ML-
76555-584EA 95.5 USD InStock
76555-584
InVivoPure™ pH 8.0 Dilution Buffer, BioXCell
Buffers Biological Buffers
InVivoPure™ dilution buffers are specifically formulated and tested to satisfy the stringent requirements for in vivo applications.
They are extremely low in endotoxin, have been screened for murine pathogens, tested in animal models for toxicity and are formulated with respect to buffer composition and pH to satisfy the requirements of Bio X Cell's antibodies.
This buffer does not contain calcium, magnesium, phenol red, or preservatives such as azide.
Caution: Keep contents sterile. Open only in a biological safety cabinet.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty