Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Orion™ Lab Star PH111 pH/mV Bench Meters, Thermo Scientific

Supplier: Thermo Scientific
Orion™
LSTAR1118 LSTAR1117 LSTAR1116 LSTAR1115 LSTAR1110
76557-412EA 954.61 USD InStock
76557-412 76557-424 76557-402 76557-408 76557-416
Orion™ Lab Star PH111 pH/mV Bench Meters, Thermo Scientific
Electrochemical Meters pH and Multi-Parameter Meters
Measure pH or ORP simultaneously with mV and temperature using the Orion™ Lab Star PH111 pH/mV bench meter. To enable comprehensive measurement capabilities, measure pH in a range of -2.000 to 18.000 pH, measure ORP in a range of -2000.0 to +2000.0 mV and measure temperature in a range of -5.0 to 105.0 °C or 23.0 to 221.0 °F.

  • Budget-friendly pH meter for simplified pH testing and ORP testing
  • Large 5” backlit display makes viewing measurements easy
  • Small meter footprint saves valuable bench space in your lab
  • Date and time stamped logs support GLP compliance
  • View active calibration logs with date and time to help ensure accuracy
  • 500-point log with date and time to track critical data
  • Output data and calibration logs to a printer or computer for long-term data storage
  • Stabilizing and ready indicators identify when measurements are complete
  • Large 5” display with backlight to improve ease of viewing and recording measurements
  • Large primary pH or ORP measurement shown simultaneously with mV and temperature measurements
  • Durable membrane keypad to support rugged lab use and disinfection needs
  • Small footprint to save bench space
  • IP54 rated housing protects meter against spills, splashes and dust intrusion
  • Export data to computer or printer for long-term data storage
  • Slope display
  • Time and date
  • Optional sleep mode
  • Up to ±250 mV and ±5 ºC offset adjustments
  • Continuous, auto-read, and timed read types
  • Calibration log for active pH, ORP, temperature with time and date
  • Self-test, automatic with startup

Featuring a large, backlit display for easy viewability and small footprint to save bench space, this pH meter is an ideal option for simplified, budget-friendly lab testing. Use the onscreen stabilizing and ready indicators to help identify when pH meter and ORP meter readings are complete. The pH electrode condition icon helps ensure reliable electrode performance throughout your testing.

View active pH, ORP and temperature calibration logs with date and time stamp to ensure accurate, reproducible measurements. Record key measurements using the 500-point data log with date and time stamp. Output data logs and calibration logs to a printer or computer for long-term data storage.

Certifications: CE, cTUVus, FCC Class B, CES-001, KC, RCM.

Ordering information: Three-year waranty.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty