Would you like to log in again?
Compact Glove Boxes, Yamato
Supplier: Yamato
SG-848 SG-828 SG-860 SG-835
76557-484EA 20544.82 USD InStock
76557-484 76557-482 76557-488 76557-480
Compact Glove Boxes, Yamato
Glove Boxes and Glove Bags
Compact, completely sealed glove box with transfer chamber and flat access door for basic isolation and/or containment.
  • High visibility interior, cleanliness, safety, complete containment, portability, and a draft-free atmopshere
  • Completed systems, nothing needs to be added except gas of choice: nitrogen, argon, or other inert gases
  • Installed purging valves are ideal for lowering oxygen and humidity levels
Very useful when working with toxic substances, asbestos, fibers, sewage residue, and harmful liquid vapors. Made in the USA.
Certifications: CE certification.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty