Would you like to log in again?
Sequential Sterilization Wraps, Halyard
Supplier: Halyard
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
68648 68154 10736 10830 68672 68172 68645 68636 68245 68124 10840 10745 10845 68654 10748 68640 10715 68136 68036 10718 68054 10854 68072 68018 10740 68254 68015 10720 68118 68130 10730 10872
76557-818CS 158.13 USD InStock
76557-818 76557-838 76557-816 76557-854 76557-832 76557-874 76557-830 76557-852 76557-858 76557-814 76557-836 76557-812 76557-834 76557-856 76557-862 76557-840 76557-860 76557-828 76557-866 76557-844 76557-822 76557-820 76557-864 76557-842 76557-848 76557-826 76557-868 76557-824 76557-846 76557-872 76557-850 76557-870
Sequential Sterilization Wraps, Halyard
Sterilization Wraps
The choice of sterile processing professionals, HALYARD sterilization wraps, made of strong reliable SMS non-woven fabric, have been helping keep sterilized instruments and supply sets ready for use at a moment’s notice. Ideal for use in individual medical and dental practices as well as major medical and surgery centers.
  • Industry standard for sterile processing teams around the world
  • Tear-, flame-, lint- and abrasion-resistant
  • Pre-vacuum steam sterilant penetration
  • Maintenance of package integrity (MPI) to meet FDA requirements
  • Ethylene oxide (EO) sterilant penetration and residuals
  • Class 1 flammability rating
  • Low linting
Halyard Health offers six grades of wrap fabric – from H100 to H600 to ensure precisely the right protection for your needs. Choosing is easy, the higher the number, the stronger the wrap. Halyard is also available in smaller sizes that meet the needs of smaller dental or medical offices.
POWERGUARD* technology for enhanced microbial barrier protection.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty