Would you like to log in again?
QUICK CHECK* Sterilization Wraps, Halyard
Supplier: Halyard
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
34154 37045 34184 34151 34146 34173 34149 34181 34176 34198 34182 34160 34195 34199 34175 34183 34168 34187 34163 34194 34180 34147 34161 34188 34145 34191 34158 34172 34192 34164 34186 34165 34174 34143 34157 34170 34167 34196 34148 34185 34200 34159 34162 34197 34193 34179 34190 34189 34156 34142 34171 34178
76557-938CS 277.27 USD InStock
76557-938 76557-898 76557-932 76557-976 76557-896 76557-974 76557-930 76557-936 76557-934 76557-978 76557-928 76557-926 76557-920 76557-964 76557-888 76557-962 76557-886 76557-968 76557-924 76557-966 76557-922 76557-890 76557-894 76557-972 76557-892 76557-970 76557-918 76557-916 76557-876 76557-910 76557-954 76557-952 76557-914 76557-958 76557-878 76557-956 76557-912 76557-880 76557-960 76557-884 76557-882 76557-906 76557-904 76557-948 76557-908 76557-942 76557-940 76557-946 76557-902 76557-944 76557-900 76557-950
QUICK CHECK* Sterilization Wraps, Halyard
Sterilization Wraps
Halyard QUICK CHECK sterilization wraps are the fast, easy way to ensure instrument sterility is uncompromised.
  • Two distinct, contrasting layers, translucent and opaque are bonded together making inspecting trays easier and faster than ever
  • Superior microbial barrier protection with Powerguard* technology
  • Cleared for all of the same sterilization systems as Halyard ONE-STEP Wrap
  • Made from recyclable #5 polypropylene fabric
To make specific trays stand out from the rest, simply flip the sheet and wrap with the other side out. Same protection, no protocol changes. QUICK CHECK Wraps give the same time savings of Halyard ONE-STEP Wraps, along with protection and performance you expect. Like all Halyard Wraps, QUICK CHECK provide superior microbial barrier protection with POWERGUARD technology, plus it’s cleared for all of the same sterilization systems as Halyard ONE-STEP Wrap. Because it’s ONE-STEP, it’s easy to make the switch to QUICK CHECK Wrap. So, you can enjoy the benefits of this innovative wrap without having to retrain staff or change protocols.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty